Learn More
Medchemexpress LLC Quercetin-3'-O-glucoside | 19254-30-9 | 464.38 | 5 MG
SDP
Catalog No. 5000416526
Encompass_Preferred
Quercetin-3'-O-glucoside is an orally active flavonoid glycoside with significant biological activities. It reduces liver glucose-6-phosphatase and regulates antioxidant enzyme activities. This compound inhibits DNA topoisomerase II and induces S-phase cell cycle arrest and apoptosis in hepatocellular carcinoma cells. It also inhibits EGFR-mediated signaling pathways and growth factor-induced migration in pancreatic cancer cells, alongside its free radical scavenging effects.
  • Reduces liver glucose-6-phosphatase activity
  • Regulates antioxidant enzyme activities
  • Inhibits DNA topoisomerase II
  • Induces S-phase cell cycle arrest and apoptosis in hepatocellular carcinoma cells
  • Inhibits EGFR-mediated signaling pathways
  • Exerts free radical scavenging effects
Catalog No. 50-004-16526 Supplier Medchemexpress LLC Supplier No. HYN124455MG
Quercetin-3'-O-glucoside is an orally active flavonoid glycoside with significant biological activities. It reduces liver glucose-6-phosphatase and regulates antioxidant enzyme activities. This compound inhibits DNA topoisomerase II and induces S-phase cell cycle arrest and apoptosis in hepatocellular carcinoma cells. It also inhibits EGFR-mediated signaling pathways and growth factor-induced migration in pancreatic cancer cells, alongside its free radical scavenging effects.
  • Reduces liver glucose-6-phosphatase activity
  • Regulates antioxidant enzyme activities
  • Inhibits DNA topoisomerase II
  • Induces S-phase cell cycle arrest and apoptosis in hepatocellular carcinoma cells
  • Inhibits EGFR-mediated signaling pathways
  • Exerts free radical scavenging effects