Learn More
Cayman Chemical Quercetin 3-O-glucuronide, Purity: ≥98%, CAS: 22688-79-5, 25 mg
SDP
Catalog No. 502985270
Encompass_Preferred
A pharmacologically active flavonol glucuronide; reduced rat immobility time in the forced swim test (0.6 mg/kg); reduced stress-induced hyperthermia in mice (1.2 mg/kg)
Catalog No. 50-298-5270 Supplier Cayman Chemical Supplier No. 2128925 MG
A pharmacologically active flavonol glucuronide; reduced rat immobility time in the forced swim test (0.6 mg/kg); reduced stress-induced hyperthermia in mice (1.2 mg/kg)