Learn More
Cayman Chemical Quercetin 3-O-sophoroside, Purity: ≥98%, CAS: 18609-17-1, 10 mg
SDP
Catalog No. 502938519
Encompass_Preferred
A flavonoid glycoside; has a relative antioxidant capacity of 1.45 compared to Trolox in an ABTS assay; inhibits lipid peroxidation in a cell-free assay using phospholipid liposomes (IC50 = 9.2 μM)
Catalog No. 50-293-8519 Supplier Cayman Chemical Supplier No. 2859210 MG
A flavonoid glycoside; has a relative antioxidant capacity of 1.45 compared to Trolox in an ABTS assay; inhibits lipid peroxidation in a cell-free assay using phospholipid liposomes (IC50 = 9.2 μM)