Learn More
Cayman Chemical Quercetin 3-O-β-D-Glucose-7-O-β-D-Gentiobioside, Purity: ≥98%, CAS: 60778-02-1, 50 mg
SDP
Catalog No. 502911723
Encompass_Preferred
A flavonoid glycoside with lipid-lowering activity; reduces sodium oleate-induced increases in triglyceride levels in HepG2 cells at 30 µM
Catalog No. 50-291-1723 Supplier Cayman Chemical Supplier No. 3613050 MG
A flavonoid glycoside with lipid-lowering activity; reduces sodium oleate-induced increases in triglyceride levels in HepG2 cells at 30 µM