Learn More
Medchemexpress LLC HY-18085 5g , Quercetin CAS:117-39-5 Purity:98%
SDP
Catalog No. 501492188
Encompass_Preferred
Medchemexpress, HY-18085 5g Quercetin CAS:117-39-5 Formula:C15H10O7 IC50: 2.4±0.6 μM (PI3K γ), 3.0±0.0 μM (PI3K δ), 5.4±0.3 μM (PI3K β) Purity:98% Quercetin, a natural flavonoid, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC 50 of 2.4±0.6 μM, 3.0±0.0 μM and 5.4±0.3 μM for PI3K γ, PI3K δ and PI3K β, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2188 Supplier Medchemexpress LLC Supplier No. HY180855G
Medchemexpress, HY-18085 5g Quercetin CAS:117-39-5 Formula:C15H10O7 IC50: 2.4±0.6 μM (PI3K γ), 3.0±0.0 μM (PI3K δ), 5.4±0.3 μM (PI3K β) Purity:98% Quercetin, a natural flavonoid, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC 50 of 2.4±0.6 μM, 3.0±0.0 μM and 5.4±0.3 μM for PI3K γ, PI3K δ and PI3K β, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.