Learn More
Medchemexpress LLC HY-18085 500mg , Quercetin CAS:117-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18085500MG
Encompass_Preferred
HY-18085 500mg Quercetin CAS: 117-39-5 Quercetin, a natural flavonoid, is a stimulator of recombinant SIRT1 and also a PI3K inhibitor with IC50 of 2.4 μM, 3.0 μM and 5.4 μM for PI3K γ, PI3K δ and PI3K β, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9051