Learn More
Medchemexpress LLC Quercetin-7-O-β-D-glucopyranoside | 491-50-9 | 464.38 | 5 MG
SDP
Catalog No. 5000388964
Encompass_Preferred
Quercimeritrin is an orally active α-glucosidase inhibitor and a P-gp substrate, demonstrating anti-angiogenic and antioxidant activities. It effectively regulates postprandial blood glucose and exhibits significant anti-angiogenic activity, inhibiting endothelial cell proliferation and microvascular growth, suggesting its potential in diabetes and breast cancer research.
  • Orally active α-glucosidase inhibitor
  • P-gp substrate with anti-angiogenic and antioxidant activities
  • Regulates postprandial blood glucose
  • Inhibits endothelial cell proliferation and microvascular growth
  • Potential utility in diabetes and breast cancer research
Catalog No. 50-003-88964 Supplier Medchemexpress LLC Supplier No. HYN04195MG
Quercimeritrin is an orally active α-glucosidase inhibitor and a P-gp substrate, demonstrating anti-angiogenic and antioxidant activities. It effectively regulates postprandial blood glucose and exhibits significant anti-angiogenic activity, inhibiting endothelial cell proliferation and microvascular growth, suggesting its potential in diabetes and breast cancer research.
  • Orally active α-glucosidase inhibitor
  • P-gp substrate with anti-angiogenic and antioxidant activities
  • Regulates postprandial blood glucose
  • Inhibits endothelial cell proliferation and microvascular growth
  • Potential utility in diabetes and breast cancer research