Learn More
Abcam Quercetin, Antio x idative flavonoid, 100MG
Catalog No. 502707748
Encompass_Preferred
MW 302.23 Da, Purity >97%. Antioxidative flavonoid. Anticancer, antithrombotic and anti-inflammatory agent. Actions include inhibition of mitochondrial ATPase, phosphodiesterases, PI3-kinase and PIP kinase activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7748 Supplier Abcam Supplier No. AB120247100MG
MW 302.23 Da, Purity >97%. Antioxidative flavonoid. Anticancer, antithrombotic and anti-inflammatory agent. Actions include inhibition of mitochondrial ATPase, phosphodiesterases, PI3-kinase and PIP kinase activity.
The product is subject to the following: Abcam Restricted Use Statement