Learn More
AdipoGen Quercetin dihydrate
SDP
Catalog No. 502562546
Encompass_Preferred
Chemical. CAS 6151-25-3. Formula C15H10O7 . 2H20. MW 302.2 . 36.0. Isolated from Umbelliferae sp. Multipotent flavonoid antioxidant. Potent free radical scavenger. Neuroprotective. Anticancer compound with chemosensitizing activity. Cell cycle arrest, apoptosis and autophagy inducer. Proteasome inhibitor. Pleiotropic kinase inhibitor, including tyrosine protein kinase Trk, mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases, PI3-kinase activity, phospholipase A2 and protein kinase C PKC. DNA topoisomerases inhibitor. SIRT1 activator. Heat shock proteins inhibitor. Reversible fatty acid synthase inhibitor. Antithrombotic, antihistaminic and anti-inflammatory agent. Monoamine oxidase inhibitor. Vasodilatory compound. anti-diabetic compound.
Catalog No. 50-256-2546 Supplier AdipoGen Supplier No. AGCN20409G001
Chemical. CAS 6151-25-3. Formula C15H10O7 . 2H20. MW 302.2 . 36.0. Isolated from Umbelliferae sp. Multipotent flavonoid antioxidant. Potent free radical scavenger. Neuroprotective. Anticancer compound with chemosensitizing activity. Cell cycle arrest, apoptosis and autophagy inducer. Proteasome inhibitor. Pleiotropic kinase inhibitor, including tyrosine protein kinase Trk, mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases, PI3-kinase activity, phospholipase A2 and protein kinase C PKC. DNA topoisomerases inhibitor. SIRT1 activator. Heat shock proteins inhibitor. Reversible fatty acid synthase inhibitor. Antithrombotic, antihistaminic and anti-inflammatory agent. Monoamine oxidase inhibitor. Vasodilatory compound. anti-diabetic compound.