Learn More
Medchemexpress LLC HY-N0146 500mg Medchemexpress, Quercetin (dihydrate) CAS:6151-25-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0146500MG
Encompass_Preferred
Medchemexpress, HY-N0146 500mg Quercetin (dihydrate) CAS:6151-25-3 Quercetin dihydrate, a natural flavonoid, is a stimulator of recombinant SIRT1 and a PI3K inhibitor with IC50s of 2.4 μM, 3.0 μM and 5.4 μM for PI3K γ, PI3K δ and PI3K β, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2774