Learn More
Enzo Life Sciences Quercetin . dihydrate (5g). CAS: 6151-25-3
SDP
Supplier:  Enzo Life Sciences ALX385001G005
Encompass_Preferred
Antioxidant flavonoid. Inhibitor of mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases. Inhibitor of protein tyrosine kinases and protein kinase C (PKC). Induces apoptosis. Blocks cells at the G0/G1 interface. Activator of human deacetylase SIRT1. Reversible inhibitor of fatty acid synthase (FAS). Inhibits the production of the inflammatory mediators nitric oxide (NO), TNF-α and IL-12 in activated macrophages.Induces gastric relaxation in mouse stomach, mainly by suppressing Ca2+ influx through voltage-dependent calcium channels. Alternative name: 3,3',4',5,7-Pentahydroxyflavone . 2H2O. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol or 1M sodium hydroxide; almost insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9033