Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Quercetin hydrate

Catalog No. 33795125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-795-125G 25 g
33-795-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-795-125G Supplier Sigma Aldrich Supplier No. 33795125G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Quercetin is an antitrypanosomal and antileishmanial compound
  • Quercetin is a well-known bioflavonoid and its influence on tumour growth in vivo has been reported
  • Quercetic exhibits leishmanicidal effect on the amastigote stage of leishmania donovani.quercetin hydrate was used as leishmanicidal flavonol to investigate the inhibition of recombinant arginase from leishmania amazonensis by quercetin, quercitrin and isoquercitrin

Specifications

CAS 849061-97-8
Molecular Formula C15H10O7 -+ xH2O
Linear Formula C15H10O7 -+ xH2O
MDL Number MFCD03847906
Quantity 25 g
Molecular Weight (g/mol) 302.24 (anhydrous basis)
Percent Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.