Learn More
STEMCELL Technologies Quercetin (Hydrate), Size: 5 g
SDP
Supplier:  STEMCELL Technologies 73932
Encompass
Quercetin inhibits phosphoinositide 3-kinase isoforms (beta: IC50 = 5.4 µM; gamma: IC50 = 2.4 µM; delta: IC50 = 3.0 µM) and SRC kinases but only shows slight inhibitory activity against serine/threonine kinases ERK1/2 and p38 alpha, as well as protein kinase C (Navarro-Nunez et al.). REPROGRAMMING⦁ Stimulates glycolytic metabolism and promotes reprogramming of somatic cells (Zhang et al.).⦁ Enhances reprogramming of human primary somatic cells to induced pluripotent stem cells (Zhu et al.).CANCER RESEARCH⦁ Induces hypoxia-inducible factor-1α (HIF-1α) and inhibits cell proliferation (Triantafyllou et al.). ⦁ Promotes apoptosis of glioblastoma cells when used together with temozolomide (Sang et al.).⦁ Reduces tumorigenicity of pancreatic cancer cells (Fan et al.).
Catalog No. 50-197-6151
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov