Learn More
Indofine Chemical Quercetin With Hpl, 99%, 10 Mg
SDP
Catalog No. 50908761
Encompass_Preferred
QUERCETIN with HPLC 10 MG, Flavone, 3,5,7,3',4'-Pentahydroxyflavone, C15H10O7, MW: 302.25, 99+%, 117-39-5, MFCD00149487
Catalog No. 50-908-761 Supplier Indofine Chemical Supplier No. 021135S 10 MG
QUERCETIN with HPLC 10 MG, Flavone, 3,5,7,3',4'-Pentahydroxyflavone, C15H10O7, MW: 302.25, 99+%, 117-39-5, MFCD00149487