Learn More
TARGETMOL CHEMICALS INC QUERCIMERITRIN 5MG
SDP
Catalog No. 502578898
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Quercimeritrin (Quercetin-7-O-beta-D-glucopyranoside) has antibacterial activity it shows promising activity against Staphylococcus aureus. purity: 98%
Catalog No. 50-257-8898 Supplier TARGETMOL CHEMICALS INC Supplier No. TN10415MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Quercimeritrin (Quercetin-7-O-beta-D-glucopyranoside) has antibacterial activity it shows promising activity against Staphylococcus aureus. purity: 98%