Learn More
Medchemexpress LLC Quercitrin | 522-12-3 | 25 MG
SDP
Catalog No. 5000389532
Encompass_Preferred
Quercitrin (Quercetin 3-rhamnoside) is a bioflavonoid compound that exhibits potential anti-inflammatory, antioxidative, and neuroprotective properties. It has been observed to induce apoptosis in colon cancer cells and is suitable for research related to cardiovascular and neurological diseases.
  • Bioflavonoid compound
  • Potential anti-inflammatory effects
  • Antioxidative properties
  • Neuroprotective effects
  • Induces apoptosis of colon cancer cells
  • Applicable for cardiovascular disease research
  • Applicable for neurological disease research
Catalog No. 50-003-89532 Supplier Medchemexpress LLC Supplier No. 25MGHYN0418R
Quercitrin (Quercetin 3-rhamnoside) is a bioflavonoid compound that exhibits potential anti-inflammatory, antioxidative, and neuroprotective properties. It has been observed to induce apoptosis in colon cancer cells and is suitable for research related to cardiovascular and neurological diseases.
  • Bioflavonoid compound
  • Potential anti-inflammatory effects
  • Antioxidative properties
  • Neuroprotective effects
  • Induces apoptosis of colon cancer cells
  • Applicable for cardiovascular disease research
  • Applicable for neurological disease research