Learn More
Perkin Elmer US LLC Quest ATR Top Plate - Diamond, Extended Range
Catalog No. 502113269
Encompass_Preferred
The Quest ATR Accessory is a performance single-reflection ATR accessory designed for sample analysis in the mid- and far-infrared. This accessory has reflective gold-coated optics and Specac's Synopti-Focal Array technology with precision-molded aspheric mirrors. The Quest ATR Accessory is compatible with Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3269 Supplier Perkin Elmer US LLC Supplier No. L1272504
Add to Cart
Add to Cart
Request a Quote
The Quest ATR Accessory is a performance single-reflection ATR accessory designed for sample analysis in the mid- and far-infrared. This accessory has reflective gold-coated optics and Specac's Synopti-Focal Array technology with precision-molded aspheric mirrors. The Quest ATR Accessory is compatible with Spectrum 3/Frontier spectrometers.