Learn More
Perkin Elmer US LLC Quest ATR Top Plate - Ge
Catalog No. 502113127
Encompass_Preferred
The Quest ATR is compatible with the Spectrum 3/Frontier spectrometer.
Catalog No. 50-211-3127 Supplier Perkin Elmer US LLC Supplier No. L1272514
The Quest ATR is compatible with the Spectrum 3/Frontier spectrometer.