Learn More
Med Vet International Zoetis QuestPlus, Equine Oral Gel, 14g Syringe
Catalog No. 502188580
Encompass_Preferred
QuestPlus (moxidectin/praziquantel) Equine Oral Gel when administered at the recommended dose level of 0.4 mg moxidectin/kg and 2.5 mg praziquantel/kg (2.2 lb) body weight treats and controls large strongyles, small strongyles, encysted cyathostomes, ascarids, pinworms, hairworms, large-mouth stomach worms, horse stomach bots, and tapeworms. Also suppresses strongyle egg production for 84 days. For horses and ponies 6 months of age or older. Safe for pregnant mares. See package insert for complete product details.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8580 Supplier Med Vet International Supplier No. PHQUEST14
QuestPlus (moxidectin/praziquantel) Equine Oral Gel when administered at the recommended dose level of 0.4 mg moxidectin/kg and 2.5 mg praziquantel/kg (2.2 lb) body weight treats and controls large strongyles, small strongyles, encysted cyathostomes, ascarids, pinworms, hairworms, large-mouth stomach worms, horse stomach bots, and tapeworms. Also suppresses strongyle egg production for 84 days. For horses and ponies 6 months of age or older. Safe for pregnant mares. See package insert for complete product details.Products are not always manufacturer specific. Actual product appearance may differ from website image.