Learn More
Cayman Chemical QuetIapIn SulfoxIde 10mg
SDP
Catalog No. 502945635
Encompass_Preferred
An inactive metabolite of quetiapine; formed from quetiapine via CYP3A
Catalog No. 50-294-5635 Supplier Cayman Chemical Supplier No. 4203510 MG
An inactive metabolite of quetiapine; formed from quetiapine via CYP3A