Learn More
Cayman Chemical Quetiapine Sulfoxide, Purity: ≥95%, CAS: 329216-63-9, 5 mg
SDP
Catalog No. 502945125
Encompass_Preferred
An inactive metabolite of quetiapine; formed from quetiapine via CYP3A
Catalog No. 50-294-5125 Supplier Cayman Chemical Supplier No. 420355 MG
An inactive metabolite of quetiapine; formed from quetiapine via CYP3A