Learn More
Medchemexpress LLC HY-14544 10mg , Quetiapine CAS:111974-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1454410MG
Encompass_Preferred
HY-14544 10mg Quetiapine CAS: 111974-69-7 Quetiapine (ICI204636) is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0866