Learn More
Medchemexpress LLC Quetiapine-d4 hemifumarate | 1217310-65-0 | 99.8% | 445.57 | C21H21D4N3O2 · 1/2C4H4O4 | 10 MG
SDP
Catalog No. 5000298895
Encompass_Preferred
Quetiapine-d4 hemifumarate is the deuterium-labeled form of quetiapine supplied as a hemifumarate salt for analytical and research use, primarily as an internal standard for LC-MS quantitation and method development.
  • Deuterium-labeled quetiapine for use as an analytical internal standard.
  • High purity suitable for quantitative analysis (99.8%).
  • Supplied as the hemifumarate salt to improve stability and handling.
  • Molecular weight 445.57 and CAS number 1217310-65-0 for precise identification.
  • Available in small measured sizes for assay development, including 10 MG packages.
Catalog No. 50-002-98895 Supplier Medchemexpress LLC Supplier No. HYB0031S110MG
Quetiapine-d4 hemifumarate is the deuterium-labeled form of quetiapine supplied as a hemifumarate salt for analytical and research use, primarily as an internal standard for LC-MS quantitation and method development.
  • Deuterium-labeled quetiapine for use as an analytical internal standard.
  • High purity suitable for quantitative analysis (99.8%).
  • Supplied as the hemifumarate salt to improve stability and handling.
  • Molecular weight 445.57 and CAS number 1217310-65-0 for precise identification.
  • Available in small measured sizes for assay development, including 10 MG packages.