Learn More
Medchemexpress LLC Quetiapine-d4 hemifumarate | 1217310-65-0 | 99.8% | C21H21D4N3O2S.1/2C4H4O4 | 1 MG
SDP
Catalog No. 5000433571
Encompass_Preferred
Quetiapine-d4 hemifumarate is the deuterium-labeled form of Quetiapine hemifumarate. This compound functions as a 5-HT receptors agonist and a dopamine receptor antagonist, exhibiting antidepressant and anxiolytic effects.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A and 5-HT2A receptors
  • Isotope-labeled for research applications
Catalog No. 50-004-33571 Supplier Medchemexpress LLC Supplier No. HYB0031S11MG
Quetiapine-d4 hemifumarate is the deuterium-labeled form of Quetiapine hemifumarate. This compound functions as a 5-HT receptors agonist and a dopamine receptor antagonist, exhibiting antidepressant and anxiolytic effects.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets 5-HT1A and 5-HT2A receptors
  • Isotope-labeled for research applications