Learn More
Medchemexpress LLC Quetiapine-d4 hemifumarate | 1217310-65-0 | 99.8% | C21H21D4N3O2S.1/2C4H4O4 | 5 MG
SDP
Catalog No. 5000432368
Encompass_Preferred
Quetiapine-d4 hemifumarate is the deuterium labeled Quetiapine hemifumarate. It acts as a 5-HT receptors agonist and a dopamine receptor antagonist, exhibiting antidepressant and anxiolytic effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32368 Supplier Medchemexpress LLC Supplier No. HYB0031S15MG
Quetiapine-d4 hemifumarate is the deuterium labeled Quetiapine hemifumarate. It acts as a 5-HT receptors agonist and a dopamine receptor antagonist, exhibiting antidepressant and anxiolytic effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.