Learn More
Apexbio Technology LLC QUETIAPINE-D8-FUMARATE-1MG
SDP
Catalog No. 5000570212
Encompass_Preferred
Quetiapine-d8 fumarate (CAS No 1185247-12-4) is a deuterium-labeled derivative of quetiapine fumarate commonly used in biomedical research for pharmacokinetic studies and drug metabolism investigations Originating from the antipsychotic family it functions by antagonizing neurotransmitter receptors such as serotonin 5-HT2A and dopamine D2 influencing neurotransmitter pathways and synaptic transmission Its in vitro activity typically ranges from nanomolar to low micromolar concentrations reflecting its effective binding at target sites Common applications include use in cell culture models animal studies and high-throughput drug screening where it helps elucidate the pharmacodynamics and pharmacokinetics of quetiapine Experimental concentrations typically depend on the specific research design allowing for tailored investigations to ascertain its therapeutic and side effect profiles
Catalog No. 50-005-70212 Supplier Apexbio Technology LLC Supplier No. SL10421
Quetiapine-d8 fumarate (CAS No 1185247-12-4) is a deuterium-labeled derivative of quetiapine fumarate commonly used in biomedical research for pharmacokinetic studies and drug metabolism investigations Originating from the antipsychotic family it functions by antagonizing neurotransmitter receptors such as serotonin 5-HT2A and dopamine D2 influencing neurotransmitter pathways and synaptic transmission Its in vitro activity typically ranges from nanomolar to low micromolar concentrations reflecting its effective binding at target sites Common applications include use in cell culture models animal studies and high-throughput drug screening where it helps elucidate the pharmacodynamics and pharmacokinetics of quetiapine Experimental concentrations typically depend on the specific research design allowing for tailored investigations to ascertain its therapeutic and side effect profiles