Learn More
Apexbio Technology LLC QUETIAPINE D8 HEMIFUM-1mg
SDP
Catalog No. 5000897632
Encompass_Preferred
Quetiapine-d8 Hemifumarate (piperazine-d8) (CAS No 1435938-24-1) is a deuterated derivative of Quetiapine which is commonly used in biomedical research for its antipsychotic properties Originating from the piperazine class it primarily targets dopamine D2 and serotonin 5-HT2A receptors modulating neurotransmitter pathways to exert its effects This compound exhibits activity in the nanomolar to low micromolar range making it suitable for in vitro studies Quetiapine-d8 Hemifumarate is often applied in cellular models and animal studies to investigate psychiatric disorders and evaluate drug metabolism and pharmacokinetics in preclinical trials It is also useful in drug screening processes providing insights into therapeutic efficacy and safety Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97632 Supplier Apexbio Technology LLC Supplier No. SL10401
Quetiapine-d8 Hemifumarate (piperazine-d8) (CAS No 1435938-24-1) is a deuterated derivative of Quetiapine which is commonly used in biomedical research for its antipsychotic properties Originating from the piperazine class it primarily targets dopamine D2 and serotonin 5-HT2A receptors modulating neurotransmitter pathways to exert its effects This compound exhibits activity in the nanomolar to low micromolar range making it suitable for in vitro studies Quetiapine-d8 Hemifumarate is often applied in cellular models and animal studies to investigate psychiatric disorders and evaluate drug metabolism and pharmacokinetics in preclinical trials It is also useful in drug screening processes providing insights into therapeutic efficacy and safety Experimental concentrations typically depend on the specific research design