Learn More
Apexbio Technology LLC Quetiapine Fumarate 111974-72-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090508
Encompass_Preferred
Quetiapine Fumarate (CAS 111974-72-2) is a small-molecule atypical antipsychotic with antagonistic activity at dopamine (D1 D2) serotonin (5-HT1A 5-HT2) and adrenergic ( 1 2) receptors In vitro it exhibits affinity for multiple neurotransmitter receptors with dopamine-2 binding characteristics similar to clozapine Preclinical in vivo studies demonstrate antipsychotic effects minimal extrapyramidal side effects and a lower propensity for sustained prolactin elevation Quetiapine has been shown to modulate neurotrophin expression and restore hippocampal neurogenesis following chronic stress in animal models supporting its value in neuropsychiatric and neuropharmacological research
Catalog No. 50-000-90508 Supplier Apexbio Technology LLC Supplier No. B14905.1
Quetiapine Fumarate (CAS 111974-72-2) is a small-molecule atypical antipsychotic with antagonistic activity at dopamine (D1 D2) serotonin (5-HT1A 5-HT2) and adrenergic ( 1 2) receptors In vitro it exhibits affinity for multiple neurotransmitter receptors with dopamine-2 binding characteristics similar to clozapine Preclinical in vivo studies demonstrate antipsychotic effects minimal extrapyramidal side effects and a lower propensity for sustained prolactin elevation Quetiapine has been shown to modulate neurotrophin expression and restore hippocampal neurogenesis following chronic stress in animal models supporting its value in neuropsychiatric and neuropharmacological research