Learn More
Selleck Chemical LLC Quetiapine Fumarate 25mg 111974-72-2 ICI-204636
SDP
Supplier:  Selleck Chemical LLC S176325MG
Encompass_Preferred
Quetiapine Fumarate (ICI-204636) is an atypical antipsychotic used in the treatment of schizophrenia, bipolar I mania, bipolar II depression, bipolar I depression and shows affinity for various neurotransmitter receptors including serotonin, dopamine, histamine, and adrenergic receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-533-7