Learn More
Selleck Chemical LLC Quetiapine Fumarate 100mg 111974-72-2 ICI-204636
SDP
Catalog No. 505339
Encompass_Preferred
Quetiapine Fumarate (ICI-204636) is an atypical antipsychotic used in the treatment of schizophrenia, bipolar I mania, bipolar II depression, bipolar I depression and shows affinity for various neurotransmitter receptors including serotonin, dopamine, histamine, and adrenergic receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-533-9 Supplier Selleck Chemical LLC Supplier No. S1763100MG
Quetiapine Fumarate (ICI-204636) is an atypical antipsychotic used in the treatment of schizophrenia, bipolar I mania, bipolar II depression, bipolar I depression and shows affinity for various neurotransmitter receptors including serotonin, dopamine, histamine, and adrenergic receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.