Learn More
Medchemexpress LLC Quetiapine sulfoxide dihydrochloride | 329218-11-3 | MFCD09031385 | 98.6% | 472.43 g·mol-1 | C21H27Cl2N3O3S | 10 MG
SDP
Catalog No. 5000297501
Encompass_Preferred
Quetiapine sulfoxide dihydrochloride is the dihydrochloride salt of quetiapine sulfoxide, a primary metabolite of the antipsychotic quetiapine. It is provided as a research-grade biochemical reference for analytical, metabolic, and pharmacokinetic studies.
  • Main metabolite of quetiapine used in metabolism research and metabolite identification.
  • Provided as a stable dihydrochloride salt to improve solubility and handling.
  • High purity suitable for analytical and reference-standard applications.
  • Available in small milligram pack sizes and as a 10 mM solution for assay preparation.
Catalog No. 50-002-97501 Supplier Medchemexpress LLC Supplier No. HYG0014A10MG
Quetiapine sulfoxide dihydrochloride is the dihydrochloride salt of quetiapine sulfoxide, a primary metabolite of the antipsychotic quetiapine. It is provided as a research-grade biochemical reference for analytical, metabolic, and pharmacokinetic studies.
  • Main metabolite of quetiapine used in metabolism research and metabolite identification.
  • Provided as a stable dihydrochloride salt to improve solubility and handling.
  • High purity suitable for analytical and reference-standard applications.
  • Available in small milligram pack sizes and as a 10 mM solution for assay preparation.