Learn More
Electron Microscopy Sciences Quetol 651/NSA Embedding Kit
SDP
Catalog No. 50980428
Encompass
A water miscible epoxy resin used for both light and electron microscopy. Its low viscosity permits easy infiltration and embedding, and yields light-colored blocks. Kit consists of: 225 ml Quetol 651, 450 ml NMA , 450 ml NSA, 50 ml
Catalog No. 50-980-428 Supplier Electron Microscopy Sciences Supplier No. 14640
A water miscible epoxy resin used for both light and electron microscopy. Its low viscosity permits easy infiltration and embedding, and yields light-colored blocks. Kit consists of: 225 ml Quetol 651, 450 ml NMA , 450 ml NSA, 50 ml