Learn More
Medchemexpress LLC Queuine dihydrochloride | 86496-18-6 | 97.3% | 350.20 g·mol-1 | C12H17Cl2N5O3 | 10 MG
SDP
Catalog No. 5000295919
Encompass_Preferred
Queuine dihydrochloride is the dihydrochloride salt of queuine, a modified nucleobase that serves as a substrate for tRNA guanine transglycosylase (TGT). It is used in research to study tRNA modification, mitochondrial function, and metabolic regulation in models of cancer and autoimmune disease.
  • Selective substrate for tRNA guanine transglycosylase (TGT).
  • Facilitates study of tRNA modification and effects on mitochondrial function and metabolism.
  • Useful in research on cancer metabolism and autoimmune disease models.
  • Available in small research quantities (e.g. 10 MG) for laboratory experiments.
  • Reported purity approximately 97.3% and molecular weight 350.20 g·mol-1.
Catalog No. 50-002-95919 Supplier Medchemexpress LLC Supplier No. HYN10574A10MG
Queuine dihydrochloride is the dihydrochloride salt of queuine, a modified nucleobase that serves as a substrate for tRNA guanine transglycosylase (TGT). It is used in research to study tRNA modification, mitochondrial function, and metabolic regulation in models of cancer and autoimmune disease.
  • Selective substrate for tRNA guanine transglycosylase (TGT).
  • Facilitates study of tRNA modification and effects on mitochondrial function and metabolism.
  • Useful in research on cancer metabolism and autoimmune disease models.
  • Available in small research quantities (e.g. 10 MG) for laboratory experiments.
  • Reported purity approximately 97.3% and molecular weight 350.20 g·mol-1.