Learn More
Zymo Research Corporation Quick-16S™ NGS Library Prep Kit, 96 rxns
SDP
Catalog No. NC1621444
Encompass_Preferred
The Quick-16S NGS Library Prep Kit utilizes real-time (quantitative) PCR (qPCR) rather than endpoint PCR for 16S rRNA amplification, enabling direct quantification of PCR products and eliminating the need for additional library quantification analysis such as TapeStation analysis or gel electrophoresis. An enzymatic clean-up is introduced between the two PCR steps, saving time and reducing costs as compared to lengthy AMPure bead-based clean-ups. With these features, the kit dramatically reduces the hands-on time of 16S library preparation. 
Catalog No. NC1621444 Supplier Zymo Research Corporation Supplier No. D6400
Add to Cart
Add to Cart
The Quick-16S NGS Library Prep Kit utilizes real-time (quantitative) PCR (qPCR) rather than endpoint PCR for 16S rRNA amplification, enabling direct quantification of PCR products and eliminating the need for additional library quantification analysis such as TapeStation analysis or gel electrophoresis. An enzymatic clean-up is introduced between the two PCR steps, saving time and reducing costs as compared to lengthy AMPure bead-based clean-ups. With these features, the kit dramatically reduces the hands-on time of 16S library preparation. 