Learn More
Zymo Research Corporation Quick-16S Plus NGS Library Prep Kit (V4) (96 preps) with Index Primer Set 2
SDP
Catalog No. NC2233807
Encompass_Preferred
The Quick-16S Plus NGS Library Prep Kit (V4) is the fastest and simplest NGS library prep targeting the V4 region of the 16S rRNA gene for high-throughput sequencing. The automation-friendly protocol utilizes a premixed amplification plate and a single qPCR/PCR for combined targeted amplification and barcode addition. Pooling by equal volume and a single clean-up of the final library, rather than massive AMPure bead-based clean-ups, simplifies the library prep process. Additional library quantification analysis such as TapeStation analysis or gel electrophoresis are not necessary. With these features, the workflow dramatically reduces the hands-on time of library preparation to only 30 minutes.
This item is not returnable. View return policy
Catalog No. NC2233807 Supplier Zymo Research Corporation Supplier No. D6430PS2
The Quick-16S Plus NGS Library Prep Kit (V4) is the fastest and simplest NGS library prep targeting the V4 region of the 16S rRNA gene for high-throughput sequencing. The automation-friendly protocol utilizes a premixed amplification plate and a single qPCR/PCR for combined targeted amplification and barcode addition. Pooling by equal volume and a single clean-up of the final library, rather than massive AMPure bead-based clean-ups, simplifies the library prep process. Additional library quantification analysis such as TapeStation analysis or gel electrophoresis are not necessary. With these features, the workflow dramatically reduces the hands-on time of library preparation to only 30 minutes.