Learn More
Zymo Research Corporation Quick-16S Primer Set V1-V2, 96 rxns
SDP
Catalog No. 502037177
Encompass_Preferred
Component for Quick-16S NGS Library Prep Kit. The primers contain adapters that are fully compatible with Illumina Nextera Index Kit or similar.
Catalog No. 50-203-7177 Supplier Zymo Research Corporation Supplier No. D64051400
Component for Quick-16S NGS Library Prep Kit. The primers contain adapters that are fully compatible with Illumina Nextera Index Kit or similar.