Learn More
Zymo Research Corporation Quick-16S™ Primer Set V3-V4, 400 µl
SDP
Catalog No. 502037178
Encompass_Preferred
Component for Quick-16S NGS Library Prep Kit. The primers contain adapters that are fully compatible with Illumina Nextera Index Kit or similar.
Catalog No. 50-203-7178 Supplier Zymo Research Corporation Supplier No. D64052400
Component for Quick-16S NGS Library Prep Kit. The primers contain adapters that are fully compatible with Illumina Nextera Index Kit or similar.