Learn More
INDUSTRIAL TEST SYSTEMS INC Quick Arsenic II Test Kit 50 tests.
SDP
Catalog No. NC3038507
Encompass_Preferred
Quick™ Arsenic II Test Kit 50 tests. For Water Analysis. Performance & accuracy verified by USEPA-ETV (www.epa.gov/etv)• Ideal for field testing requiring no technical training• Used by International Aid Organizations for field testing• Results in only 12-14 minutes using safe, convenient reagents• Components clearly labeled and color coded for error free testing• 99.8% Correlation to ICP-MS reference method
Catalog No. NC3038507 Supplier INDUSTRIAL TEST SYSTEMS INC Supplier No. 481303
Quick™ Arsenic II Test Kit 50 tests. For Water Analysis. Performance & accuracy verified by USEPA-ETV (www.epa.gov/etv)• Ideal for field testing requiring no technical training• Used by International Aid Organizations for field testing• Results in only 12-14 minutes using safe, convenient reagents• Components clearly labeled and color coded for error free testing• 99.8% Correlation to ICP-MS reference method