Learn More
Mti Corporation QUICK ASSY SPLIT COIN CELL
SDP
Catalog No. NC1386259
Encompass
Quick Assembly Split Coin Cell for Battery Material testing, No Crimper Needed
Catalog No. NC1386259 Supplier Mti Corporation Supplier No. HSTC20
Quick Assembly Split Coin Cell for Battery Material testing, No Crimper Needed