Learn More
Electron Microscopy Sciences Glass Knife Quick Boat for 6.0 mm Knife
SDP
Catalog No. 50930664
Encompass_Preferred
These boats are made from aluminum and are anodized black. They come complete with a set screw on one side. The area where the glass knife will be attached, is cut to fit to the width of the knife and is cushioned by silicone rubber to ensure a tight seal and no leakage.
Catalog No. 50-930-664 Supplier Electron Microscopy Sciences Supplier No. 7100960
These boats are made from aluminum and are anodized black. They come complete with a set screw on one side. The area where the glass knife will be attached, is cut to fit to the width of the knife and is cushioned by silicone rubber to ensure a tight seal and no leakage.