Learn More
Electron Microscopy Sciences Quick-Bond - Aron Alpha 200 5 x 2g/bx
SDP
Catalog No. 50949013
Encompass_Preferred
Aron Alpha, Type 200, is a one component, room temperature curing structural adhesive composed mainly of alpha cyanoacrylate monomer in a liquid state. When this monomer is applied in a single thin coating to the bonding surface of the material, it instantly solidifies through polymerization to produce an extra high strength, colorless bond with virtually no shrinkage.
Catalog No. 50-949-013 Supplier Electron Microscopy Sciences Supplier No. 72588
Aron Alpha, Type 200, is a one component, room temperature curing structural adhesive composed mainly of alpha cyanoacrylate monomer in a liquid state. When this monomer is applied in a single thin coating to the bonding surface of the material, it instantly solidifies through polymerization to produce an extra high strength, colorless bond with virtually no shrinkage.