Learn More
HANNA INSTRUMENTS INC Quick Calibration Solution (500mL Bottle)
SDP
Supplier:  HANNA INSTRUMENTS INC HI982825
Encompass
HI9828-25 is a quick calibration solution for use with Hanna's family of portable multiparameter meters. It provides for a single point of calibration for pH and conductivity and is ideal for field use.Single, Field Ready CalibrationLeak Proof Portable Solution
Catalog No. NC1734248