Learn More
Zymo Research Corporation Quick-cfDNA™ Serum & Plasma Kit (50 Preps)
SDP
Catalog No. NC0984867
Encompass_Preferred
A combination of chemical and enzymatic methods are used to efficiently recover total DNA (including cell-free apoptotic, necrotic, mitochondrial and viral DNA) linearly from up to 10 ml of sample. Zymo-Spin™ technology allows for ultra-pure DNA to be eluted in as little as 35 l water. The resulting DNA is suitable for all subsequent analyses and molecular manipulations such as qPCR, Next-Generation sequencing and DNA methylation analyses. Zymos serum and plasma cell-free DNA extraction technology will empower your discovery of circulating DNA biomarkers.
Catalog No. NC0984867 Supplier Zymo Research Corporation Supplier No. D4076
A combination of chemical and enzymatic methods are used to efficiently recover total DNA (including cell-free apoptotic, necrotic, mitochondrial and viral DNA) linearly from up to 10 ml of sample. Zymo-Spin™ technology allows for ultra-pure DNA to be eluted in as little as 35 l water. The resulting DNA is suitable for all subsequent analyses and molecular manipulations such as qPCR, Next-Generation sequencing and DNA methylation analyses. Zymos serum and plasma cell-free DNA extraction technology will empower your discovery of circulating DNA biomarkers.