Learn More
Zymo Research Corporation Quick-cfDNA/cfRNA Serum & Plasma Kit
SDP
Supplier:  Zymo Research Corporation R1072
Encompass_Preferred
The kit does not use toxic phenol-chloroform or cumbersome protein precipitation and removal steps. The kit efficiently separates cell-free DNA and RNA, with option to co-elute. Cell-free nucleic acids scale linearly relative to the sample input volume up to 3 ml. Zymo-Spin™ Technology enables isolation of ultra-pure cell-free nucleic acids suitable for all subsequent analyses, such as RT-qPCR and NGS.
Catalog No. 50-195-7598