Learn More
Zymo Research Corporation Quick-cfRNA Serum & Plasma Kit
SDP
Catalog No. 501251684
Encompass_Preferred
The Quick-cfRNA Serum & Plasma Kit enables simple, reliable, and efficient preparation of high-quality circulating cell-free RNA (including protein-bound, exosomal, miRNA and other small RNAs) from serum, plasma and other biological fluids. The kit does not use toxic phenol-chloroform or cumbersome protein precipitation and removal steps. Recovered cell-free RNA scales linearly relative to the sample input volume up to 3 ml. ZymoSpin™ Technology enables isolation of ultra-pure cell-free RNA suitable for all subsequent analyses, such as RT-qPCR and NGS. Empower your discovery of circulating RNA biomarkers.
Catalog No. 50-125-1684 Supplier Zymo Research Corporation Supplier No. R1059
The Quick-cfRNA Serum & Plasma Kit enables simple, reliable, and efficient preparation of high-quality circulating cell-free RNA (including protein-bound, exosomal, miRNA and other small RNAs) from serum, plasma and other biological fluids. The kit does not use toxic phenol-chloroform or cumbersome protein precipitation and removal steps. Recovered cell-free RNA scales linearly relative to the sample input volume up to 3 ml. ZymoSpin™ Technology enables isolation of ultra-pure cell-free RNA suitable for all subsequent analyses, such as RT-qPCR and NGS. Empower your discovery of circulating RNA biomarkers.