Learn More
TA Instruments Quick Change Disposable Plate Holder, Required for Use with Aluminum Quick Change Disposable Plates, 8 mm 20 mm 25 mm, with P/N 402619.901 Standard Flat Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580834 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Quick Change Disposable Plate Holder is required when using HR X0, DHR, ARES-G2, APS, Aluminum Quick Change Disposable Plates (8 mm, 20 mm, 25 mm) with P/N 402619.901 ARES-G2, APS, Standard Flat Plate.
Catalog No. 50-005-80834 Supplier TA Instruments Supplier No. 402751.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS, Quick Change Disposable Plate Holder is required when using HR X0, DHR, ARES-G2, APS, Aluminum Quick Change Disposable Plates (8 mm, 20 mm, 25 mm) with P/N 402619.901 ARES-G2, APS, Standard Flat Plate.