Learn More
Grainger Micro100 Locking Screw, 1/4-28 Holder Thread, Uncoated, 10/pk
Catalog No. 502776654
Encompass_Preferred
This locking screw is designed for use with compatible quick change holders, providing a secure and reliable fastening solution for your machining setup. Engineered for precision and durability, this component ensures optimal performance in tool retention applications.
  • Features a 1/4-28 holder thread
  • Designed with an M6 screw diameter size
  • Uncoated finish for standard applications
  • Compatible with QDS and QDSM manufacturer models
Catalog No. 50-277-6654 Supplier Grainger Supplier No. 18D325
This locking screw is designed for use with compatible quick change holders, providing a secure and reliable fastening solution for your machining setup. Engineered for precision and durability, this component ensures optimal performance in tool retention applications.
  • Features a 1/4-28 holder thread
  • Designed with an M6 screw diameter size
  • Uncoated finish for standard applications
  • Compatible with QDS and QDSM manufacturer models