Learn More
Brady Quick Charger for M410 Printer Label Printer
Catalog No. 17467576 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System
Catalog No. For Use With (Equipment)
17-467-576 BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System
1 options
Catalog No. 17-467-576 Supplier Brady Supplier No. M410QC
Add to Cart
Add to Cart
This Quick Charger charges the rechargeable NiMH battery in the M410 Label Printer faster than the standard AC Adaptor.
  • Package includes the Quick Charger only (printer and battery sold separately).
  • With a quick charger on hand, you can be sure your battery is ready to go when you need it most.
  • Power supply formatted for North America.
  • Plugs directly into battery and charges battery from a wall outlet rather than from inside the printer.
Specifications
Product Type Quick Charger
For Use With (Equipment) BMP41 Label Printer, Brady M410 Label Printer, BMP61 Labeling System