Learn More
COLDER PRODUCTS COMPANY QUICK CNNCT CPLNG INSRT25PK
Catalog No. 503131426
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 50 mm 1 98 in Gray with Dark Gray Overmold For Use With 9 5mm Includes Shut-off valves Polypropylene Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm
Catalog No. 50-313-1426 Supplier COLDER PRODUCTS COMPANY Supplier No. NS4D22006
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 50 mm 1 98 in Gray with Dark Gray Overmold For Use With 9 5mm Includes Shut-off valves Polypropylene Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm