Learn More
Angio Proteomie CCP1679 Quick Coating Solution, CAP, 0.1%
SDP
Catalog No. NC0731668
Encompass
The CCP1679 Quick Coating Solution from Angioproteomie is a ready-to-use coating solution for ELISA plates and other labware. It is composed of carbonate-bicarbonate buffer and contains no proteins. The solution has a pH of 9.6 and is supplied in bottles containing 100 ml. It can coat 96 wells of an ELISA plate in just 5 minutes. The CCP1679 allows rapid and uniform coating of plates for immunoassays. It is a convenient reagent that eliminates weighing and mixing steps.
Catalog No. NC0731668 Supplier Angio Proteomie Supplier No. CAP01
The CCP1679 Quick Coating Solution from Angioproteomie is a ready-to-use coating solution for ELISA plates and other labware. It is composed of carbonate-bicarbonate buffer and contains no proteins. The solution has a pH of 9.6 and is supplied in bottles containing 100 ml. It can coat 96 wells of an ELISA plate in just 5 minutes. The CCP1679 allows rapid and uniform coating of plates for immunoassays. It is a convenient reagent that eliminates weighing and mixing steps.